Product Details

SNP ID
rs199505178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63521516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGGGACCTCTCCTCTCCAGGCCG[C/T]CTGGGTTCCACTTCCAGCAACAGCT
Phenotype
MIM: 602004
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPDPF PubMed Links

Gene Details

Gene
PPDPF
Gene Name
pancreatic progenitor cell differentiation and proliferation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024299.2 200 Silent Mutation NP_077275.1
Gene
PTK6
Gene Name
protein tyrosine kinase 6
There are no transcripts associated with this gene.

View Full Product Details