Product Details

SNP ID
rs199517242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37112350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAATCTGCAGATTGTTCCTGCTT[A/C]AGTAGGTTTGATGTGCCAGGGATCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MROH8 PubMed Links

Gene Details

Gene
MROH8
Gene Name
maestro heat like repeat family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152503.5 2926 Missense Mutation NP_689716.4
NM_213631.2 2926 Intron NP_998796.1
NM_213632.2 2926 Intron NP_998797.2

View Full Product Details