Product Details

SNP ID
rs199534402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44750381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCCTCGGGGCCGTCGCCTTCT[C/T]GCACTTCGAGGGCTGGACCTTCTTC
Phenotype
MIM: 607368 MIM: 611601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNK15 PubMed Links

Gene Details

Gene
KCNK15
Gene Name
potassium two pore domain channel subfamily K member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022358.3 600 Missense Mutation TCG,TTG S179L NP_071753.2
XM_017028003.1 600 Intron XP_016883492.1
Gene
KCNK15-AS1
Gene Name
KCNK15 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
RIMS4
Gene Name
regulating synaptic membrane exocytosis 4
There are no transcripts associated with this gene.

View Full Product Details