Product Details

SNP ID
rs202009773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46161985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCGTCACGCCCGGGAACACGTA[A/G]GCCAGGATGCGGCGGTCCAGCTGGA
Phenotype
MIM: 606806 MIM: 612412
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FTCD PubMed Links

Gene Details

Gene
FTCD
Gene Name
formimidoyltransferase cyclodeaminase
There are no transcripts associated with this gene.

Gene
SPATC1L
Gene Name
spermatogenesis and centriole associated 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142854.1 366 Silent Mutation NP_001136326.1
NM_032261.4 366 Silent Mutation NP_115637.3
XM_005261188.4 366 Silent Mutation XP_005261245.1
XM_011529756.2 366 Silent Mutation XP_011528058.1
XM_017028480.1 366 Silent Mutation XP_016883969.1

View Full Product Details