Product Details

SNP ID
rs201632967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44612186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGTGACGTGGGCCATGTCAGCC[A/G]AGTCTCCTCCCCCAGCACCTGCACT
Phenotype
MIM: 612920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP10-8 PubMed Links

Gene Details

Gene
KRTAP10-8
Gene Name
keratin associated protein 10-8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198695.2 108 Missense Mutation CAA,CGA Q29R NP_941968.2
Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 108 Intron NP_001258966.1
NM_144991.2 108 Intron NP_659428.2

View Full Product Details