Product Details

SNP ID
rs202030727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44124707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAGGTTCGAGATCTCTTGCAAC[C/T]TGTCTTTGGACGCCATGGAGGTGAG
Phenotype
MIM: 601659 MIM: 601475
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C21orf33 PubMed Links

Gene Details

Gene
C21orf33
Gene Name
chromosome 21 open reading frame 33
There are no transcripts associated with this gene.

Gene
PWP2
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005049.2 Intron NP_005040.2
XM_011529667.2 Intron XP_011527969.1

View Full Product Details