Product Details

SNP ID
rs201238667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32311696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTGTTCACAGGAACAGCCCCAA[A/G]CACCACCAAACATGCCCCTGGAGTC
Phenotype
MIM: 609196 MIM: 608865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105372779 PubMed Links

Gene Details

Gene
LOC105372779
Gene Name
uncharacterized LOC105372779
There are no transcripts associated with this gene.

Gene
MRAP
Gene Name
melanocortin 2 receptor accessory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285394.1 290 Silent Mutation AAA,AAG K14K NP_001272323.1
NM_178817.3 290 Silent Mutation AAA,AAG K73K NP_848932.1
NM_206898.1 290 Intron NP_996781.1
XM_006724028.3 290 Silent Mutation AAA,AAG K73K XP_006724091.1
XM_017028407.1 290 Intron XP_016883896.1
Gene
URB1
Gene Name
URB1 ribosome biogenesis 1 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014825.2 290 UTR 3 NP_055640.2

View Full Product Details