Product Details

SNP ID
rs200563007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:36135249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGGCCAACAGGGGCATCGGCTT[G/T]GCCATCGCGCGCGAACTGTGCCGAC
Phenotype
MIM: 603608
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CBR3 PubMed Links

Gene Details

Gene
CBR3
Gene Name
carbonyl reductase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001236.3 349 Missense Mutation TTG,TTT L19F NP_001227.1
XM_011529772.2 349 Missense Mutation TTG,TTT L19F XP_011528074.1
Gene
CBR3-AS1
Gene Name
CBR3 antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC100133286
Gene Name
uncharacterized LOC100133286
There are no transcripts associated with this gene.

View Full Product Details