Product Details

SNP ID
rs200646454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:14373790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGAACTCTTGAATGACTTGAC[A/G]GATCCGAGGAATACGAGTGGAGCCC
Phenotype
MIM: 601100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPA13 PubMed Links

Gene Details

Gene
HSPA13
Gene Name
heat shock protein family A (Hsp70) member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006948.4 1312 Missense Mutation CGT,TGT R415C NP_008879.3

View Full Product Details