Product Details

SNP ID
rs200489704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30592674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGCTGTGGCTACAGAGGCCTG[G/T]ACTGTGGCTATGGCTGTGGCTATGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP22-1 PubMed Links

Gene Details

Gene
KRTAP22-1
Gene Name
keratin associated protein 22-1
There are no transcripts associated with this gene.

Gene
KRTAP22-2
Gene Name
keratin associated protein 22-2
There are no transcripts associated with this gene.

Gene
KRTAP6-2
Gene Name
keratin associated protein 6-2
There are no transcripts associated with this gene.

Gene
KRTAP6-3
Gene Name
keratin associated protein 6-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181605.3 235 Missense Mutation GAC,TAC D77Y NP_853636.3

View Full Product Details