Product Details
- SNP ID
-
rs200155645
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:38086245 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAGGGCGGGCAAGGGCTCACCTT[C/G]ACGTCAGTGGCAACTGCTGTGCTGC
- Phenotype
-
MIM: 610409
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BAIAP2L2
PubMed Links
Gene Details
- Gene
- BAIAP2L2
- Gene Name
- BAI1 associated protein 2 like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025045.5 |
1890 |
Silent Mutation |
GTC,GTG |
V488V |
NP_079321.3 |
XM_005261751.4 |
1890 |
Silent Mutation |
GTC,GTG |
V488V |
XP_005261808.1 |
XM_011530379.2 |
1890 |
Silent Mutation |
GTC,GTG |
V474V |
XP_011528681.1 |
XM_011530380.2 |
1890 |
Silent Mutation |
GTC,GTG |
V470V |
XP_011528682.1 |
XM_011530381.2 |
1890 |
Silent Mutation |
GTC,GTG |
V469V |
XP_011528683.1 |
XM_011530382.2 |
1890 |
Silent Mutation |
GTC,GTG |
V469V |
XP_011528684.1 |
XM_011530383.2 |
1890 |
Silent Mutation |
GTC,GTG |
V464V |
XP_011528685.1 |
XM_011530384.2 |
1890 |
Silent Mutation |
GTC,GTG |
V458V |
XP_011528686.1 |
XM_011530386.2 |
1890 |
Silent Mutation |
GTC,GTG |
V293V |
XP_011528688.1 |
XM_011530387.2 |
1890 |
Silent Mutation |
GTC,GTG |
V293V |
XP_011528689.1 |
XM_011530388.2 |
1890 |
Silent Mutation |
GTC,GTG |
V281V |
XP_011528690.1 |
- Gene
- SLC16A8
- Gene Name
- solute carrier family 16 member 8
There are no transcripts associated with this gene.
View Full Product Details