Product Details

SNP ID
rs200418035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30287166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCGGCCCTCACCATGCTGAGTG[C/T]GGGGAAAGCCATTGCTGGAATCATC
Phenotype
MIM: 617034 MIM: 610020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GATSL3 PubMed Links

Gene Details

Gene
GATSL3
Gene Name
GATS protein like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037666.2 624 Missense Mutation CAC,CGC H165R NP_001032755.1
Gene
TBC1D10A
Gene Name
TBC1 domain family member 10A
There are no transcripts associated with this gene.

View Full Product Details