Product Details

SNP ID
rs201658679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36141537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGATGGCACGGATTTCACTCCCA[A/G]TGGTTTGTGTGGCTTCGTAATAATT
Phenotype
MIM: 607253
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOL3 PubMed Links

Gene Details

Gene
APOL3
Gene Name
apolipoprotein L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014349.2 1843 Missense Mutation ACT,ATT T220I NP_055164.1
NM_030644.1 1843 Missense Mutation ACT,ATT T220I NP_085147.1
NM_145639.1 1843 Missense Mutation ACT,ATT T220I NP_663614.1
NM_145640.2 1843 Missense Mutation ACT,ATT T291I NP_663615.1
NM_145641.2 1843 Missense Mutation ACT,ATT T91I NP_663616.1
NM_145642.2 1843 Missense Mutation ACT,ATT T91I NP_663617.1
XM_006724324.1 1843 Missense Mutation ACT,ATT T220I XP_006724387.1
XM_006724325.2 1843 Missense Mutation ACT,ATT T220I XP_006724388.1
XM_017028946.1 1843 Missense Mutation ACT,ATT T221I XP_016884435.1
XM_017028947.1 1843 Missense Mutation ACT,ATT T221I XP_016884436.1
XM_017028948.1 1843 Missense Mutation ACT,ATT T221I XP_016884437.1
XM_017028949.1 1843 Missense Mutation ACT,ATT T221I XP_016884438.1
XM_017028950.1 1843 Missense Mutation ACT,ATT T221I XP_016884439.1
XM_017028951.1 1843 Missense Mutation ACT,ATT T221I XP_016884440.1
XM_017028952.1 1843 Missense Mutation ACT,ATT T91I XP_016884441.1

View Full Product Details