Product Details

SNP ID
rs199561166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23180998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGCTTCGCGGAGGCGTGGAAGG[A/C]GCAGTTCCCGGACTCAGAGCCCCCG
Phenotype
MIM: 151410 MIM: 605663
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BCR PubMed Links

Gene Details

Gene
BCR
Gene Name
BCR, RhoGEF and GTPase activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004327.3 634 Missense Mutation GAG,GCG E13A NP_004318.3
NM_021574.2 634 Missense Mutation GAG,GCG E13A NP_067585.2
Gene
RSPH14
Gene Name
radial spoke head 14 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014433.2 634 Intron NP_055248.1
XM_011530149.2 634 Intron XP_011528451.1
XM_011530150.1 634 Intron XP_011528452.1
XM_011530151.2 634 Intron XP_011528453.1
XM_011530152.2 634 Intron XP_011528454.1
XM_011530154.2 634 Intron XP_011528456.1
XM_011530155.2 634 Intron XP_011528457.1
XM_017028774.1 634 Intron XP_016884263.1

View Full Product Details