Product Details

SNP ID
rs199524344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20112623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTATCTGGGGGTCCTGGTGTGGG[C/T]TGAGCTGGAGGGCTGTGGGGCCCCA
Phenotype
MIM: 609030 MIM: 601180 MIM: 611151
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGCR8 PubMed Links

Gene Details

Gene
DGCR8
Gene Name
DGCR8 microprocessor complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190326.1 2068 Intron NP_001177255.1
NM_022720.6 2068 Intron NP_073557.3
XM_006724268.2 2068 Intron XP_006724331.1
Gene
MIR6816
Gene Name
microRNA 6816
There are no transcripts associated with this gene.

Gene
RANBP1
Gene Name
RAN binding protein 1
There are no transcripts associated with this gene.

Gene
TRMT2A
Gene Name
tRNA methyltransferase 2 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257994.1 2068 UTR 3 NP_001244923.1
NM_022727.5 2068 Silent Mutation CAA,CAG Q606Q NP_073564.3
NM_182984.4 2068 Silent Mutation CAA,CAG Q606Q NP_892029.2
XM_011530139.2 2068 Silent Mutation CAA,CAG Q624Q XP_011528441.1
XM_011530141.1 2068 Silent Mutation CAA,CAG Q298Q XP_011528443.1
XM_011530142.2 2068 Silent Mutation CAA,CAG Q298Q XP_011528444.1
XM_017028770.1 2068 Silent Mutation CAA,CAG Q624Q XP_016884259.1

View Full Product Details