Product Details
- SNP ID
-
rs199859140
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:28696915 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATTTCTTACCTTTATAAGACAGT[C/G]CTCTTCTTGAGATGACAGTAAAACA
- Phenotype
-
MIM: 604373
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CHEK2
PubMed Links
Gene Details
- Gene
- CHEK2
- Gene Name
- checkpoint kinase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005735.1 |
973 |
Missense Mutation |
CAC,GAC |
H404D |
NP_001005735.1 |
NM_001257387.1 |
973 |
Missense Mutation |
CAC,GAC |
H140D |
NP_001244316.1 |
NM_007194.3 |
973 |
Missense Mutation |
CAC,GAC |
H361D |
NP_009125.1 |
NM_145862.2 |
973 |
Intron |
|
|
NP_665861.1 |
XM_006724114.3 |
973 |
Missense Mutation |
CAC,GAC |
H212D |
XP_006724177.2 |
XM_006724116.2 |
973 |
Missense Mutation |
CAC,GAC |
H180D |
XP_006724179.2 |
XM_011529839.2 |
973 |
Missense Mutation |
CAC,GAC |
H414D |
XP_011528141.1 |
XM_011529840.2 |
973 |
Intron |
|
|
XP_011528142.1 |
XM_011529841.1 |
973 |
Missense Mutation |
CAC,GAC |
H337D |
XP_011528143.1 |
XM_011529842.2 |
973 |
Missense Mutation |
CAC,GAC |
H304D |
XP_011528144.1 |
XM_011529843.1 |
973 |
Missense Mutation |
CAC,GAC |
H294D |
XP_011528145.1 |
XM_011529844.2 |
973 |
Intron |
|
|
XP_011528146.1 |
XM_011529845.2 |
973 |
Missense Mutation |
CAC,GAC |
H140D |
XP_011528147.1 |
XM_017028560.1 |
973 |
Missense Mutation |
CAC,GAC |
H402D |
XP_016884049.1 |
XM_017028561.1 |
973 |
Missense Mutation |
CAC,GAC |
H140D |
XP_016884050.1 |
View Full Product Details