Product Details

SNP ID
rs201300764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42585854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGCCCCTGAGGACTTGAAGAG[C/T]GCCTTCAGGATGGTGTCAGGGTCCA
Phenotype
MIM: 611520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLDIP3 PubMed Links

Gene Details

Gene
POLDIP3
Gene Name
DNA polymerase delta interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278657.1 1308 Silent Mutation GCA,GCG A418A NP_001265586.1
NM_032311.4 1308 Silent Mutation GCA,GCG A401A NP_115687.2
NM_178136.2 1308 Silent Mutation GCA,GCG A372A NP_835237.1
XM_011530457.1 1308 UTR 3 XP_011528759.1
XM_017029010.1 1308 UTR 3 XP_016884499.1
XM_017029011.1 1308 Intron XP_016884500.1
Gene
RRP7BP
Gene Name
ribosomal RNA processing 7 homolog B, pseudogene
There are no transcripts associated with this gene.

View Full Product Details