Product Details

SNP ID
rs201945163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37944446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATTTCAGCAAGGATGATTCCTC[A/G]GTACTGTTTGCCCTGTCCCAGGGCC
Phenotype
MIM: 604414
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf23 PubMed Links

Gene Details

Gene
C22orf23
Gene Name
chromosome 22 open reading frame 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207062.1 606 Nonsense Mutation CGA,TGA R164* NP_001193991.1
NM_032561.4 606 Nonsense Mutation CGA,TGA R185* NP_115950.3
XM_005261781.1 606 Nonsense Mutation CGA,TGA R185* XP_005261838.1
XM_005261782.3 606 Nonsense Mutation CGA,TGA R185* XP_005261839.1
XM_005261783.2 606 Nonsense Mutation CGA,TGA R164* XP_005261840.1
XM_005261784.2 606 Nonsense Mutation CGA,TGA R164* XP_005261841.1
Gene
MICALL1
Gene Name
MICAL like 1
There are no transcripts associated with this gene.

Gene
POLR2F
Gene Name
RNA polymerase II subunit F
There are no transcripts associated with this gene.

View Full Product Details