Product Details

SNP ID
rs199679384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21000697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTTCAGCGGGGCCAGGTGGGTA[A/T]CTGTGTCCTTTGGTCTTGGTTGTCC
Phenotype
MIM: 600574 MIM: 608077 MIM: 609518
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LZTR1 PubMed Links

Gene Details

Gene
LZTR1
Gene Name
leucine zipper like transcription regulator 1
There are no transcripts associated with this gene.

Gene
P2RX6
Gene Name
purinergic receptor P2X 6
There are no transcripts associated with this gene.

Gene
THAP7
Gene Name
THAP domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008695.1 531 Missense Mutation AGA,AGT R109S NP_001008695.1
NM_030573.2 531 Missense Mutation AGA,AGT R109S NP_085050.2
Gene
THAP7-AS1
Gene Name
THAP7 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TUBA3FP
Gene Name
tubulin alpha 3f pseudogene
There are no transcripts associated with this gene.

View Full Product Details