Product Details

SNP ID
rs199810262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29769697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAGAAGCTGTGGAAACACATCA[A/G]GCACAAGTATGAGAACAAGTAGTTC
Phenotype
MIM: 610843
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UQCR10 PubMed Links

Gene Details

Gene
UQCR10
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003684.1 259 UTR 3 NP_001003684.1
NM_013387.3 259 Missense Mutation AAG,AGG K57R NP_037519.2
Gene
ZMAT5
Gene Name
zinc finger matrin-type 5
There are no transcripts associated with this gene.

View Full Product Details