Product Details

SNP ID
rs200016238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43168025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGTGCCTAGACAAGGCAGGTAA[C/T]GTGGCAGCCACCGGGCTGGTCCAGA
Phenotype
MIM: 109610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSPO PubMed Links

Gene Details

Gene
TSPO
Gene Name
translocator protein
There are no transcripts associated with this gene.

Gene
TTLL12
Gene Name
tubulin tyrosine ligase like 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015140.3 1980 Missense Mutation ATT,GTT I640V NP_055955.1

View Full Product Details