Product Details

SNP ID
rs201264983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232528278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTCCAGGGCTGGACAGACAACC[A/G]GCTGAAGTGGAATGCTGAAGAATTT
Phenotype
MIM: 100720 MIM: 613858
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRND PubMed Links

Gene Details

Gene
CHRND
Gene Name
cholinergic receptor nicotinic delta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000751.2 288 Missense Mutation CAG,CGG Q87R NP_000742.1
NM_001256657.1 288 Missense Mutation CAG,CGG Q72R NP_001243586.1
NM_001311195.1 288 UTR 5 NP_001298124.1
NM_001311196.1 288 UTR 5 NP_001298125.1
XM_011510524.2 288 UTR 5 XP_011508826.1
Gene
PRSS56
Gene Name
protease, serine 56
There are no transcripts associated with this gene.

View Full Product Details