Product Details

SNP ID
rs201214237
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231593393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCTGAGAGAGAGCCGGAGTCA[A/G]CCCCCTCTGCCCCGAGTGCTGAGCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C2orf57 PubMed Links

Gene Details

Gene
C2orf57
Gene Name
chromosome 2 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152614.2 530 Missense Mutation ACC,GCC T148A NP_689827.2

View Full Product Details