Product Details

SNP ID
rs201066011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86449895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGGAGAGCATTTTTAGTAGAAC[A/G]TAATTTGGTTTTAGCTGAACGAAAG
Phenotype
MIM: 611512
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM3A PubMed Links

Gene Details

Gene
KDM3A
Gene Name
lysine demethylase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146688.1 333 Missense Mutation CAT,CGT H92R NP_001140160.1
NM_018433.5 333 Missense Mutation CAT,CGT H92R NP_060903.2
XM_006712051.3 333 UTR 5 XP_006712114.1
XM_017004493.1 333 Missense Mutation CAT,CGT H92R XP_016859982.1
XM_017004494.1 333 UTR 5 XP_016859983.1

View Full Product Details