Product Details

SNP ID
rs201081009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99322209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGATGTCTGTCTAGTATTTTACA[A/C]CTGTAAGTGACCACACATAGAAAAT
Phenotype
MIM: 612564
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TXNDC9 PubMed Links

Gene Details

Gene
TXNDC9
Gene Name
thioredoxin domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005783.3 459 Missense Mutation AGG,AGT R103S NP_005774.2
XM_017003147.1 459 Missense Mutation AGG,AGT R103S XP_016858636.1

View Full Product Details