Product Details

SNP ID
rs200653506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200490331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCGTGCCTCCATTTTCAGCGCT[C/T]GCAGCCCCAGGGGAAGCCTGAGGAT
Phenotype
MIM: 613817
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD18 PubMed Links

Gene Details

Gene
KCTD18
Gene Name
potassium channel tetramerization domain containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321547.1 1592 Silent Mutation GCA,GCG A350A NP_001308476.1
NM_001321548.1 1592 Silent Mutation GCA,GCG A141A NP_001308477.1
NM_001321550.1 1592 Silent Mutation GCA,GCG A141A NP_001308479.1
NM_152387.3 1592 Silent Mutation GCA,GCG A350A NP_689600.2
Gene
SPATS2L
Gene Name
spermatogenesis associated serine rich 2 like
There are no transcripts associated with this gene.

View Full Product Details