Product Details

SNP ID
rs201084315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219547557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCACTGGGTTGCTGTTTGGCT[C/T]GGTGGTCATCTTCCTCCTCTGCTAC
Phenotype
MIM: 606715 MIM: 610405 MIM: 610991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASIC4 PubMed Links

Gene Details

Gene
ASIC4
Gene Name
acid sensing ion channel subunit family member 4
There are no transcripts associated with this gene.

Gene
CHPF
Gene Name
chondroitin polymerizing factor
There are no transcripts associated with this gene.

Gene
MIR3132
Gene Name
microRNA 3132
There are no transcripts associated with this gene.

Gene
OBSL1
Gene Name
obscurin like 1
There are no transcripts associated with this gene.

Gene
TMEM198
Gene Name
transmembrane protein 198
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005209.2 629 Missense Mutation TCG,TTG S73L NP_001005209.1
NM_001303098.1 629 Missense Mutation TCG,TTG S73L NP_001290027.1
XM_017003367.1 629 Missense Mutation TCG,TTG S73L XP_016858856.1

View Full Product Details