Product Details

SNP ID
rs201245165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219544752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCCGGGGACTGTGGCAACACTG[C/T]GGTTCCAGCTGCTGCCCCCTGAGCC
Phenotype
MIM: 606715 MIM: 610405 MIM: 610991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASIC4 PubMed Links

Gene Details

Gene
ASIC4
Gene Name
acid sensing ion channel subunit family member 4
There are no transcripts associated with this gene.

Gene
CHPF
Gene Name
chondroitin polymerizing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195731.1 436 Intron NP_001182660.1
NM_024536.5 436 Intron NP_078812.2
XM_011511838.2 436 Intron XP_011510140.1
Gene
MIR3132
Gene Name
microRNA 3132
There are no transcripts associated with this gene.

Gene
OBSL1
Gene Name
obscurin like 1
There are no transcripts associated with this gene.

Gene
TMEM198
Gene Name
transmembrane protein 198
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005209.2 436 Missense Mutation CGG,TGG R9W NP_001005209.1
NM_001303098.1 436 Missense Mutation CGG,TGG R9W NP_001290027.1
XM_017003367.1 436 Missense Mutation CGG,TGG R9W XP_016858856.1

View Full Product Details