Product Details

SNP ID
rs200948491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241814524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCCTTCACGGACAGTGCTCTTC[G/T]AGCGGGAGAGGACGGGCCTGACCTA
Phenotype
MIM: 608527
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC107986004 PubMed Links

Gene Details

Gene
LOC107986004
Gene Name
uncharacterized LOC107986004
There are no transcripts associated with this gene.

Gene
NEU4
Gene Name
neuraminidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167599.2 754 Missense Mutation GAG,TAG E27* NP_001161071.1
NM_001167600.2 754 Missense Mutation GAG,TAG E14* NP_001161072.1
NM_001167601.2 754 Missense Mutation GAG,TAG E14* NP_001161073.1
NM_001167602.2 754 Missense Mutation GAG,TAG E14* NP_001161074.1
NM_080741.3 754 Missense Mutation GAG,TAG E26* NP_542779.2

View Full Product Details