Product Details

SNP ID
rs200632383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232407921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCGCCTACGCCCACACGGTGAA[C/T]CGCAACTGGTACTCGGATGCCGACG
Phenotype
MIM: 171810
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALPPL2 PubMed Links

Gene Details

Gene
ALPPL2
Gene Name
alkaline phosphatase, placental like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031313.2 Intron NP_112603.2

View Full Product Details