Product Details

SNP ID
rs199625082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73258990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCCCCCTACCCGGGTTAGCAGC[C/T]GCCTTCCACCTTGATGTGCAGAATC
Phenotype
MIM: 605140 MIM: 609108
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCT7 PubMed Links

Gene Details

Gene
CCT7
Gene Name
chaperonin containing TCP1 subunit 7
There are no transcripts associated with this gene.

Gene
FBXO41
Gene Name
F-box protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080410.2 3604 Missense Mutation AGC,GGC S874G NP_001073879.2
XM_005264158.3 3604 Missense Mutation AGC,GGC S480G XP_005264215.1
XM_006711950.2 3604 Missense Mutation AGC,GGC S874G XP_006712013.1
XM_011532562.1 3604 Missense Mutation AGC,GGC S874G XP_011530864.1
XM_011532563.2 3604 Missense Mutation AGC,GGC S626G XP_011530865.1
XM_017003421.1 3604 Missense Mutation AGC,GGC S874G XP_016858910.1
XM_017003422.1 3604 Missense Mutation AGC,GGC S874G XP_016858911.1
XM_017003423.1 3604 Missense Mutation AGC,GGC S874G XP_016858912.1

View Full Product Details