Product Details

SNP ID
rs199560829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:45006397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGTCTGGCGTCCCCGATGGAG[C/T]CTTCTCATCCTCCGAGCTGCCTAAC
Phenotype
MIM: 604994
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIX2 PubMed Links

Gene Details

Gene
SIX2
Gene Name
SIX homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016932.4 942 Missense Mutation ACT,GCT T217A NP_058628.3
XM_005264100.3 942 Missense Mutation ACT,GCT T219A XP_005264157.1

View Full Product Details