Product Details

SNP ID
rs199952966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240029683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAAGCAAGGCTGTATAGAAGAC[G/T]GTGACCACGGTGAGGTGAGAGGCGC
Phenotype
MIM: 603835
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NDUFA10 PubMed Links

Gene Details

Gene
NDUFA10
Gene Name
NADH:ubiquinone oxidoreductase subunit A10
There are no transcripts associated with this gene.

Gene
OR6B2
Gene Name
olfactory receptor family 6 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005853.1 747 Silent Mutation NP_001005853.1

View Full Product Details