Product Details
- SNP ID
-
rs199722039
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:200490386 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTGCCCCCGTGCCCACCAGGGCC[A/G]TGGCTCTGGAAGGTGCAGAGCGCTG
- Phenotype
-
MIM: 613817
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCTD18
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs13018579] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCTD18
- Gene Name
- potassium channel tetramerization domain containing 18
- Gene
- SPATS2L
- Gene Name
- spermatogenesis associated serine rich 2 like
There are no transcripts associated with this gene.
View Full Product Details