Product Details

SNP ID
rs199763633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:19353561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAAGGCGGGCAGCTGGAAGCGC[A/G]CATCCACCAAGCTGGACACCGTGCC
Phenotype
MIM: 608891
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4757 PubMed Links

Gene Details

Gene
MIR4757
Gene Name
microRNA 4757
There are no transcripts associated with this gene.

Gene
OSR1
Gene Name
odd-skipped related transciption factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145260.2 1417 Missense Mutation NP_660303.1
XM_006711942.3 1417 Missense Mutation XP_006712005.1

View Full Product Details