Product Details

SNP ID
rs200537408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241814577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGCCCTCGCTGCTCCCCGTGCC[C/G]CCCGGGCCCACCCTGCTGGCCTTTG
Phenotype
MIM: 608527
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC107986004 PubMed Links

Gene Details

Gene
LOC107986004
Gene Name
uncharacterized LOC107986004
There are no transcripts associated with this gene.

Gene
NEU4
Gene Name
neuraminidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167599.2 807 Silent Mutation CCC,CCG P44P NP_001161071.1
NM_001167600.2 807 Silent Mutation CCC,CCG P31P NP_001161072.1
NM_001167601.2 807 Silent Mutation CCC,CCG P31P NP_001161073.1
NM_001167602.2 807 Silent Mutation CCC,CCG P31P NP_001161074.1
NM_080741.3 807 Silent Mutation CCC,CCG P43P NP_542779.2

View Full Product Details