Product Details

SNP ID
rs199738000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:8731066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTCAGGGCTGCTGGCTATCAAG[C/T]TGCATTCGGAAGGGCTTTTCTTGTC
Phenotype
MIM: 615759
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIDINS220 PubMed Links

Gene Details

Gene
KIDINS220
Gene Name
kinase D-interacting substrate 220kDa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020738.2 5152 Missense Mutation AAC,AGC N1657S NP_065789.1

View Full Product Details