Product Details

SNP ID
rs200129430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218992167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGCTGCTGCTGCCACTCCAGG[C/T]GCTCCAGCGAGGATAGTCTCCCTTC
Phenotype
MIM: 600836 MIM: 607150 MIM: 611173
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYBA2 PubMed Links

Gene Details

Gene
CRYBA2
Gene Name
crystallin beta A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057093.1 273 Missense Mutation ACC,GCC T80A NP_476434.1
NM_057094.1 273 Missense Mutation ACC,GCC T80A NP_476435.1
Gene
FEV
Gene Name
FEV, ETS transcription factor
There are no transcripts associated with this gene.

Gene
MIR375
Gene Name
microRNA 375
There are no transcripts associated with this gene.

View Full Product Details