Product Details

SNP ID
rs200493515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:153478473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCGTCGGTCACCACGGACGCCTG[A/G]GTGCAGCAGCGCACGGCTCGCTCCA
Phenotype
MIM: 612171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPRM PubMed Links

Gene Details

Gene
RPRM
Gene Name
reprimo, TP53 dependent G2 arrest mediator candidate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019845.2 336 Silent Mutation ACC,ACT T31T NP_062819.1

View Full Product Details