Product Details

SNP ID
rs200311995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24203682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCAAAACGGACCCAGACCTCCC[C/T]GGTGGGGACCTCATGCAGCAGCAGT
Phenotype
MIM: 604464
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITSN2 PubMed Links

Gene Details

Gene
ITSN2
Gene Name
intersectin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006277.2 5215 Missense Mutation AGG,GGG R1680G NP_006268.2
NM_019595.3 5215 Missense Mutation AGG,GGG R1653G NP_062541.3
NM_147152.2 5215 Intron NP_671494.2

View Full Product Details