Product Details

SNP ID
rs199769227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105242181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACTTTGGGGACCACTTCTGCC[A/G]CCTCTCAGCCACGCTCTACTGGTTT
Phenotype
MIM: 604838 MIM: 606237
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR45 PubMed Links

Gene Details

Gene
GPR45
Gene Name
G protein-coupled receptor 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007227.3 439 Missense Mutation CAC,CGC H108R NP_009158.3
Gene
TGFBRAP1
Gene Name
transforming growth factor beta receptor associated protein 1
There are no transcripts associated with this gene.

View Full Product Details