Product Details

SNP ID
rs200063708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27377566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTCAGGATGAGTCTGTGTGGAC[A/G]GCCCGGCTGCCAGCAGTCCCCAGGG
Phenotype
MIM: 606687 MIM: 605119 MIM: 605963 MIM: 613598
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2B4 PubMed Links

Gene Details

Gene
EIF2B4
Gene Name
eukaryotic translation initiation factor 2B subunit delta
There are no transcripts associated with this gene.

Gene
PPM1G
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1G
There are no transcripts associated with this gene.

Gene
SNX17
Gene Name
sorting nexin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267059.1 1746 Intron NP_001253988.1
NM_001267060.1 1746 Intron NP_001253989.1
NM_001267061.1 1746 Intron NP_001253990.1
NM_014748.3 1746 Intron NP_055563.1
XM_011533203.1 1746 Intron XP_011531505.1
XM_017005405.1 1746 Intron XP_016860894.1
Gene
ZNF513
Gene Name
zinc finger protein 513
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201459.1 1746 Silent Mutation GCC,GCT A473A NP_001188388.1
NM_144631.5 1746 Silent Mutation GCC,GCT A535A NP_653232.3
XM_005264142.2 1746 Silent Mutation GCC,GCT A473A XP_005264199.1
XM_005264143.3 1746 Silent Mutation GCC,GCT A367A XP_005264200.1

View Full Product Details