Product Details

SNP ID
rs200175007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85134030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCGCAGGCGGAGAGGCGCCC[A/G]CAGCCCGTCCGGGACACTTTCCAGA
Phenotype
MIM: 604652
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCF7L1 PubMed Links

Gene Details

Gene
TCF7L1
Gene Name
transcription factor 7 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031283.2 489 Silent Mutation CCA,CCG P88P NP_112573.1
XM_006712109.2 489 Silent Mutation CCA,CCG P88P XP_006712172.1

View Full Product Details