Product Details
- SNP ID
-
rs201669454
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:60948110 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCACGTACTGTGTCTCCATGAAGT[A/G]AATGACGCGAGCTCGCACAGCCAGG
- Phenotype
-
MIM: 612787
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PUS10
PubMed Links
Gene Details
- Gene
- PUS10
- Gene Name
- pseudouridylate synthase 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001322123.1 |
1523 |
Missense Mutation |
CAC,TAC |
H462Y |
NP_001309052.1 |
NM_001322124.1 |
1523 |
Missense Mutation |
CAC,TAC |
H462Y |
NP_001309053.1 |
NM_001322127.1 |
1523 |
Missense Mutation |
CAC,TAC |
H239Y |
NP_001309056.1 |
NM_144709.3 |
1523 |
Missense Mutation |
CAC,TAC |
H462Y |
NP_653310.2 |
XM_011532568.2 |
1523 |
Missense Mutation |
CAC,TAC |
H462Y |
XP_011530870.1 |
XM_011532570.1 |
1523 |
Missense Mutation |
CAC,TAC |
H462Y |
XP_011530872.1 |
XM_011532571.1 |
1523 |
Missense Mutation |
CAC,TAC |
H462Y |
XP_011530873.1 |
XM_011532572.1 |
1523 |
Missense Mutation |
CAC,TAC |
H462Y |
XP_011530874.1 |
XM_011532573.2 |
1523 |
Missense Mutation |
CAC,TAC |
H462Y |
XP_011530875.1 |
XM_011532574.2 |
1523 |
Missense Mutation |
CAC,TAC |
H462Y |
XP_011530876.1 |
XM_011532576.2 |
1523 |
Missense Mutation |
CAC,TAC |
H462Y |
XP_011530878.1 |
XM_011532578.1 |
1523 |
Missense Mutation |
CAC,TAC |
H399Y |
XP_011530880.1 |
XM_017003428.1 |
1523 |
Missense Mutation |
CAC,TAC |
H377Y |
XP_016858917.1 |
XM_017003429.1 |
1523 |
Missense Mutation |
CAC,TAC |
H377Y |
XP_016858918.1 |
View Full Product Details