Product Details

SNP ID
rs201920487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218271445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGACCCGGGCCCTGCAAAGCCA[C/T]AGCTCCCCAGAGGGCAAGGAGGAAC
Phenotype
MIM: 603488 MIM: 610147 MIM: 609023 MIM: 610364
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AAMP PubMed Links

Gene Details

Gene
AAMP
Gene Name
angio associated migratory cell protein
There are no transcripts associated with this gene.

Gene
GPBAR1
Gene Name
G protein-coupled bile acid receptor 1
There are no transcripts associated with this gene.

Gene
MIR6513
Gene Name
microRNA 6513
There are no transcripts associated with this gene.

Gene
PNKD
Gene Name
paroxysmal nonkinesigenic dyskinesia
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077399.2 276 Intron NP_001070867.1
NM_015488.4 276 Silent Mutation CAC,CAT H44H NP_056303.3
NM_022572.4 276 Intron NP_072094.1
XM_017003771.1 276 Silent Mutation CAC,CAT H44H XP_016859260.1
XM_017003772.1 276 Intron XP_016859261.1
Gene
TMBIM1
Gene Name
transmembrane BAX inhibitor motif containing 1
There are no transcripts associated with this gene.

View Full Product Details