Product Details

SNP ID
rs201715144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20447567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGGACGAGTTCCCCGAGGTGTA[C/T]GTGCCCACCGTCTTCGAGAACTATG
Phenotype
MIM: 165370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHOB PubMed Links

Gene Details

Gene
RHOB
Gene Name
ras homolog family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004040.3 497 Silent Mutation TAC,TAT Y34Y NP_004031.1

View Full Product Details