Product Details

SNP ID
rs201855352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108382470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGCTGCCACCATCCTTGCTAGA[A/G]AAAAACTGTAAGGTAAAAAAGCAAA
Phenotype
MIM: 608357
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SULT1C4 PubMed Links

Gene Details

Gene
SULT1C4
Gene Name
sulfotransferase family 1C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321770.1 808 Intron NP_001308699.1
NM_006588.3 808 Silent Mutation GAA,GAG E127E NP_006579.2
XM_017003807.1 808 Silent Mutation GAA,GAG E37E XP_016859296.1

View Full Product Details