Product Details
- SNP ID
-
rs202051938
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:169479571 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTCACCATGTCGGTGCTGGATGC[G/T]CTTTGGGAGGATCGGGATGTCCGTT
- Phenotype
-
MIM: 603650
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BBS5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10188609] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BBS5
- Gene Name
- Bardet-Biedl syndrome 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152384.2 |
76 |
Silent Mutation |
GCG,GCT |
A6A |
NP_689597.1 |
View Full Product Details