Product Details

SNP ID
rs202149799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233034172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGATGGCCACCGGTCCATGAA[C/T]CCATGCCCCTTGTATGACGCGCAGA
Phenotype
MIM: 605528
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEU2 PubMed Links

Gene Details

Gene
NEU2
Gene Name
neuraminidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005383.2 258 Silent Mutation AAC,AAT N86N NP_005374.2

View Full Product Details