Product Details

SNP ID
rs201833579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101257880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGGAACTCTTCAAAAAGACGCCT[C/T]GCCAGATTGCACTGATGGACGTTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNOT11 PubMed Links

Gene Details

Gene
CNOT11
Gene Name
CCR4-NOT transcription complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017546.4 686 Missense Mutation CGC,TGC R202C NP_060016.3

View Full Product Details